missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver chloride, Premion™, 99.997% (metals basis)

Manufacturer:  Alfa Aesar™ 1085706

 View more versions of this product

Catalog No. AA1085706



Specifications

Specifications

Silver chloride
(metals basis)
UN3077
143.32
99.997%
5.56g/mL
2.071
1550°C
455°C
7783-90-6
MFCD00003399
14,8509
Coarse Powder
Premion™
Odorless
Light Sensitive
White
5g

Chemical Identifiers

7783-90-6
MFCD00003399
Safety and Handling

Safety and Handling

GHS H Statement
H290
May be corrosive to metals.

GHS P Statement
P234-P390-P406
Keep only in original container.
Absorb spillage to prevent material damage.
Store in corrosive resistant container with a resistant inner liner.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

RTECSNumber : VW3563000

TSCA : TSCA

Documents