missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver chloride, 99+%, extra pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 211270500

 View more versions of this product

Catalog No. AC211270500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

1550.0°C
455.0°C
Powder
99+%
AgCl
05,580; 10,347
Solubility in water: 1.93 mg/l (25°C). Other solubilities: insoluble in alcohol
[Cl-].[Ag+]
143.32
≥99%
Glass bottle
Silver chloride
7783-90-6
50 g
AgCl
MFCD00003399
15,8648
HKZLPVFGJNLROG-UHFFFAOYSA-M
silver(1+) chloride
143.32
Extra Pure

Chemical Identifiers

7783-90-6
143.32
HKZLPVFGJNLROG-UHFFFAOYSA-M
[Cl-].[Ag+]
AgCl
MFCD00003399
silver(1+) chloride
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Absorb spillage to prevent material damage.
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.

GHS Signal Word: Warning

EINECSNumber : 232-033-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only