Please login to your online account to display your discounted pricing

Silver chloride, 99.9999%, (trace metal basis), ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC194040010

 View more versions of this product

Catalog No. AC194040010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 143.32
Quantity 1g
Color White to beige to gray
Purity Grade For trace metal basis analysis
Assay Percent Range 100.00%
Chemical Name or Material Silver chloride
CAS 7783-90-6
Molecular Formula AgCl
Packaging Glass bottle
Linear Formula AgCl
Total Trace Metal Impurities 1ppm max.

Specifications

Chemical Name or Material Silver chloride
CAS 7783-90-6
Purity Grade For trace metal basis analysis
Assay Percent Range 100.00%
Molecular Formula AgCl
Linear Formula AgCl
Formula Weight 143.32