Please login to your online account to display your discounted pricing

Silver chloride, 99.9999%, (trace metal basis), ACROS Organics™

$115.53 - $115.53

Chemical Identifiers

Chemical Name or Material Silver chloride
CAS 7783-90-6
Purity Grade For trace metal basis analysis
Assay Percent Range 100.00%
Molecular Formula AgCl
Linear Formula AgCl
Formula Weight 143.32
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC194040010

  SDS
thermo fisher scientific
AC194040010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC194040050

  SDS
thermo fisher scientific
AC194040050
Glass bottle 5g Each for $115.53
Description & Specifications

Specifications

Physical Form Powder
Formula Weight 143.32
Quantity 1g
Color White to beige to gray
Purity Grade For trace metal basis analysis
Assay Percent Range 100.00%
Chemical Name or Material Silver chloride
CAS 7783-90-6
Molecular Formula AgCl
Packaging Glass bottle
Linear Formula AgCl
Total Trace Metal Impurities 1ppm max.

for ultra trace analysis