Please login to your online account to display your discounted pricing

Silver chloride, 99.9999%, (trace metal basis), ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC194040050

 View more versions of this product

Catalog No. AC194040050

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 143.32
Physical Form Powder
Quantity 5g
Color White to beige to gray
Purity Grade For trace metal basis analysis
Linear Formula AgCl
Total Trace Metal Impurities 1ppm max.
Assay Percent Range 100.00%
Chemical Name or Material Silver chloride
CAS 7783-90-6
Molecular Formula AgCl
Packaging Glass bottle

Specifications

Chemical Name or Material Silver chloride
CAS 7783-90-6
Purity Grade For trace metal basis analysis
Assay Percent Range 100.00%
Molecular Formula AgCl
Linear Formula AgCl
Formula Weight 143.32