Learn More
Silver chloride, 4.9% in H{2}O, nanoparticle dispersion, <200nm APS Number Weighted, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04728036
Description
Used as pharmaceutical intermediate and in pharmaceutical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
White | |
Nanoparticle dispersion | |
1.038 g/mL | |
MFCD00003399 | |
500 g | |
Silver chloride dispersion in water | |
HKZLPVFGJNLROG-UHFFFAOYSA-M | |
silver(1+) chloride | |
143.32 | |
Odorless | |
Light sensitive |
Silver chloride | |
7783-90-6 | |
AgCl | |
UN3082 | |
14,8509 | |
Dispersible in water. Very slightly soluble in cold water,soluble in solutions of alkali cyanides,thiosulfates,ammonium carbonates. Appreciably soluble in concentrated aqueous solutions of ammonium chloride,mercuric nitrate,and silver nitrate. | |
[Cl-].[Ag+] | |
143.32 | |
4.9% in H2O | |
<200nm APS Number Weighted | |
Liquid |
Chemical Identifiers
7783-90-6 | |
143.32 | |
HKZLPVFGJNLROG-UHFFFAOYSA-M | |
silver(1+) chloride |
AgCl | |
MFCD00003399 | |
Silver chloride dispersion in water | |
[Cl-].[Ag+] |
Safety and Handling
P234-P390
H290
DOTInformation : Transport Hazard Class: 9; Packaging Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCE, LIQUID, N.O.S.
EINECSNumber : 232-033-3
RTECSNumber : VW3563000
TSCA : Yes
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only