Learn More
Silver carbonate, 99.5% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01142014
Description
Silver carbonate is used in the production of silver powder in microelectronics, conducting polymers, and in tooth surface treatment to prevent cavities. It is also used as an antimicrobial agent for infection resistant surgical fabric materials. It is used in organic synthesis, and in biological specimen staining.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver carbonate | |
534-16-7 | |
25 g | |
CH2Ag2O3 | |
carbonic acid; silver, carbonic acid disilver | |
WIKQEUJFZPCFNJ-UHFFFAOYSA-N | |
carbonic acid;silver | |
25203940 | |
Light sensitive |
218°C (decomposition) | |
Powder | |
99.5% (metals basis) | |
MFCD00003403 | |
Insoluble in water,acetone,acetate and alcohol. Freely soluble in dilute HNO3 and ammonia. | |
C(=O)(O)O.[Ag].[Ag] | |
277.76 | |
275.75 | |
6.08 g/mL |
Chemical Identifiers
534-16-7 | |
277.76 | |
WIKQEUJFZPCFNJ-UHFFFAOYSA-N | |
25203940 | |
C(=O)(O)O.[Ag].[Ag] |
CH2Ag2O3 | |
MFCD00003403 | |
carbonic acid; silver, carbonic acid disilver | |
carbonic acid;silver |
Safety and Handling
P280-P305+P351+P338-P310
H318
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 208-590-3
RTECSNumber : FG0700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only