Please login to your online account to display your discounted pricing

Silver bromide, Premion™, 99.998% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Silver bromide
CAS 7785-23-1
Name Note Premion™
Molecular Formula AgBr
Formula Weight 187.78
MDL Number MFCD00003398
UN Number UN3077
Merck Index 14,8506
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1211006

  SDS
alfa aesar
1211006
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 187.78
Quantity 5g
Sensitivity Light Sensitive
TSCA TSCA
Color Pale Yellow
Assay 99.998%
Molecular Formula AgBr
Name Note Premion™
Merck Index 14,8506
UN Number UN3077
DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
Physical Form Crystalline
CAS 7785-23-1
Density 6.47
Odor Odorless
Melting Point 432°C
Assay Percent Notes (metals basis)
Chemical Name or Material Silver bromide
MDL Number MFCD00003398