Learn More
Silver bromide, Premion™, 99.998% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01211006
Description
Silver bromide is used in photographic film. Used in production of radiograph. It also used in infrared applications as windows and lenses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silver bromide | |
Premion™ | |
Yellow | |
5 g | |
AgBr | |
UN3077 | |
silver bromide, bromargyrite, bromyrite, nanoparticles, silver i bromide, silver bromide, premion | |
ADZWSOLPGZMUMY-UHFFFAOYSA-M | |
bromosilver | |
66199 | |
99.998% | |
Light sensitive |
432°C | |
7785-23-1 | |
Crystalline | |
(metals basis) | |
MFCD00003398 | |
14,8506 | |
soluble in alkali cyanide solutions. Partially soluble in ammonia. Insoluble in alcohol and acids. Practically insoluble in water. | |
Br[Ag] | |
187.772 | |
187.78 | |
Odorless | |
6.47 |
Chemical Identifiers
7785-23-1 | |
187.772 | |
ADZWSOLPGZMUMY-UHFFFAOYSA-M | |
66199 | |
Br[Ag] |
AgBr | |
MFCD00003398 | |
silver bromide, bromargyrite, bromyrite, nanoparticles, silver i bromide, silver bromide, premion | |
bromosilver |
Safety and Handling
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 232-076-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only