missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silver bromide, 99.9%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1142506

 View more versions of this product

Catalog No. AA1142506



Specifications

Specifications

Silver bromide
99.9%
MFCD00003398
silver bromide, silver bromide, bromargyrite, bromyrite, nanoparticles, silver i bromide, silver bromide, silver bromide, silver bromide, premion
Br[Ag]
187.772
187.78
6.47g/cm3 at 20°C
Light sensitive
5g
7785-23-1
AgBr
Pale yellow powder/flake/crystalline/beads
ADZWSOLPGZMUMY-UHFFFAOYSA-M
bromosilver
66199
Powder
Odorless
Yellow

Chemical Identifiers

7785-23-1
187.772
ADZWSOLPGZMUMY-UHFFFAOYSA-M
66199
Br[Ag]
AgBr
MFCD00003398
silver bromide, silver bromide, bromargyrite, bromyrite, nanoparticles, silver i bromide, silver bromide, silver bromide, silver bromide, premion
bromosilver
Safety and Handling

Safety and Handling

Use appropriate containment to avoid environmental contamination. This material and/or its container must be disposed of as hazardous waste.

H400-H410: Very toxic to aquatic life. Very toxic to aquatic life with long lasting effects.

DOTInformation : DOT Class: 9, Packaging Group: III

EINECSNumber : 232-076-8

Documents