Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver, brazing flux, white paste

Chemical Identifiers

Chemical Name or Material Silver, brazing flux
Name Note Active range: 540 to 980°
MDL Number MFCD01868499
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41355TB

 
alfa aesar™
41355TB
113g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA41355TD

 
alfa aesar™
41355TD
454g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 113g
Chemical Name or Material Silver, brazing flux
Name Note Active range: 540 to 980°
Physical Form Paste
MDL Number MFCD01868499
Color White
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H360-H315-H319-H335
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P261-P280-P281-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H360-H315-H319-H335
May damage fertility or the unborn child.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.