Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver, brazing alloy, flux covered, 1.6mm (0.06 in.) dia., 46cm (18 in.) long

Chemical Identifiers

Chemical Name or Material Silver, brazing Alloy
Name Note Flux covered, 1.6mm (0.06 in.) dia., 46cm (18 in.) long, Tensile Strength: 79,000 psi
MDL Number MFCD00801120
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41031KG

  SDS
alfa aesar™
41031KG
1 piece CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA41031KJ

  SDS
alfa aesar™
41031KJ
5 pieces CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Name Note Flux covered, 1.6mm (0.06 in.) dia., 46cm (18 in.) long, Tensile Strength: 79,000 psi
Chemical Name or Material Silver, brazing Alloy
Avg. Mol. Wt. or Mol. Wt. Range Ag:Cu:Zn:Sn; 56:22:17:5 wt% Nontoxic, cadmium free
TSCA TSCA
MDL Number MFCD00801120
Quantity 1 piece

  • Low temperature alloy for use on ferrous and nonferrous material; good color match for stainless material