Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver, brazing alloy, 0.8mm (0.03 in.) dia.

Chemical Identifiers

Chemical Name or Material Silver, brazing Alloy
Name Note 0.8mm (0.03 in.) dia.
MDL Number MFCD00801120
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4102610

  SDS
alfa aesar™
4102610
15g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4102615

  SDS
alfa aesar™
4102615
30g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Nontoxic, Cadmium Free
Chemical Name or Material Silver, brazing Alloy
Name Note 0.8mm (0.03 in.) dia.
Avg. Mol. Wt. or Mol. Wt. Range Ag:Cu:Zn:Sn; 56:22:17:5 wt%
Quantity 15g
MDL Number MFCD00801120
TSCA TSCA