missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver bis(trifluoromethylsulfonyl)imide

Manufacturer:  Alfa Aesar™ 4678406

 View more versions of this product

Catalog No. AA4678406



Specifications

Specifications

Silver bis (trifluoromethylsulfonyl)imide
C2AgF6NO4S2
MFCD08457644
Silver triflimide; Silver bis(trifluoromethanesulfonyl)imide
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Ag+]
388.004
388
Moisture & Light Sensitive
249°C
189114-61-2
(CF3SO2)2NAg
UN1759
HSYLTRBDKXZSGS-UHFFFAOYSA-N
silver;bis(trifluoromethylsulfonyl)azanide
131875416
Solid
Orange to White
5g

Chemical Identifiers

189114-61-2
388.004
HSYLTRBDKXZSGS-UHFFFAOYSA-N
131875416
C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Ag+]
C2AgF6NO4S2
MFCD08457644
Silver triflimide; Silver bis(trifluoromethanesulfonyl)imide
silver;bis(trifluoromethylsulfonyl)azanide
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.

SDS Documents