Please login to your online account to display your discounted pricing

Silver Arsenite , in Sulfuric Acid, LabChem™

$253.77 - $253.77

Chemical Identifiers

Chemical Name or Material Silver Arsenite
CAS 7732-18-5
CAS Max % 93.7
CAS 7761-88-8
CAS Max % 3
CAS 7664-93-9
CAS Max % 2
CAS 1327-53-3
CAS Max % 1.3
Name Note in Sulfuric Acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC224222

 
lab chem
LC224222
1L Each for $253.77
Description & Specifications

Specifications

Formula Weight 446.52
Chemical Name or Material Silver Arsenite
CAS Max % 2
CAS 1327-53-3
Identification Passes Test
CAS Max % 1.3
Name Note in Sulfuric Acid
Packaging Amber Glass
Physical Form Liquid
Solubility Soluble in water
Storage Information Room Temperature
Linear Formula Ag3AsO3
CAS 7732-18-5
UN Number UN2922
Decomposition Information Nitrogen oxides
Color Clear, colorless
Quantity 1L
Health Hazard 1 Danger
CAS Max % 93.7
Health Hazard 2 GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
CAS 7761-88-8
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Store locked up.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If in skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.
CAS Max % 3
CAS 7664-93-9
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves, protective clothing, eye protection, face protection.
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Store locked up.
Avoid release to the environment.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If in skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Collect spillage.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.