missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silver arsenate, 99.99% (metals basis)

Manufacturer:  Alfa Aesar™ 4422814

 View more versions of this product

Catalog No. AA4422814



Specifications

Specifications

Silver arsenate
(metals basis)
MFCD00046163
3ag.aso4
[O-][As](=O)([O-])[O-].[Ag+].[Ag+].[Ag+]
462.522
462.53
99.99%
Red
13510-44-6
Ag3AsO4
UN1557
IMGNYAPMSDUASV-UHFFFAOYSA-K
trisilver;trioxido(oxo)-$l^{5}-arsane
166835
Powder/Lumps
Light sensitive
25g

Chemical Identifiers

13510-44-6
462.522
IMGNYAPMSDUASV-UHFFFAOYSA-K
166835
[O-][As](=O)([O-])[O-].[Ag+].[Ag+].[Ag+]
Ag3AsO4
MFCD00046163
3ag.aso4
trisilver;trioxido(oxo)-$l^{5}-arsane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H331-H350
Toxic if swallowed.
Toxic if inhaled.
May cause cancer.

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ARSENIC COMPOUNDS, SOLID, N.O.S.

EINECSNumber : 236-841-7

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents