Please login to your online account to display your discounted pricing

Silver, 1000μg/mL (0.10% w/v), BAKER INSTRA-ANALYZED™ Reagent, J.T.Baker™

Acids and standards that are purified and characterized for use in trace element analysis in the low parts-per-billion (ppb) range

$123.83 - $132.41

Chemical Identifiers

Chemical Name or Material Silver, 1000μg/mL (0.10% w/v)
Name Note Plasma Standard
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Molecular Formula Ag
Formula Weight 107.87
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003921

 
avantor performance
577904
Poly Bottle 100mL Each for $132.41

02003922

 
avantor performance
646704
Narrow Mouth Amber Glass Bottle 150mL Each for $123.83
Description & Specifications

Specifications

Formula Weight 107.87
Chemical Name or Material Silver, 1000μg/mL (0.10% w/v)
Molecular Formula Ag
Quantity 100mL
Specific Gravity 1L = 1.00kg
Name Note Plasma Standard
Purity Grade BAKER INSTRA-ANALYZED™ Reagent
Packaging Poly Bottle