missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicone oil, for melting point and boiling point apparatuses, ACROS Organics™

Manufacturer:  Acros Organics 163850010

 View more versions of this product

Catalog No. AC163850010



Specifications

Specifications

Silicone oil
63148-62-9
Poly(dimethylsiloxane)
Oily Liquid
Authentic
1.4030 to 1.4060
0.96
500 mPa.s (25°C)
-50.0°C
300°C
For melting point and boiling point apparatuses
MFCD00132673
0.00
0.9600g/mL
Glass bottle
Solubility in water: practically insoluble.
1.5% max. (5 g, 230°C, 2 hrs)
Colorless
1kg

Chemical Identifiers

63148-62-9
0.00
MFCD00132673
Poly(dimethylsiloxane)
SDS Documents