missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicone oil, high temperature, usable temperature range: 25 to 250°C (open system) and 25 to 315°C (closed system), Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04589622

 View more versions of this product

Catalog No. AA4589622



Description

Description

Silicone oil is used in high temperature open systems, closed systems and circulating, closed loop and heat transfer baths. It is used as a high temperature fluid for laboratory research apparatus and instruments. It is also used as a calibration media in temperature controlled baths.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • High temperature open system, closed system, circulating, closed loop, and heat transfer baths
  • High temperature fluids for laboratory research apparatus and instruments
Specifications

Specifications

Silicone oil
Colorless
Liquid
C16H22O2Si2
>300°C (572°F)
diphenyl-dimethylsiloxane copolymer, polydimethyl-diphenylsiloxane, viscosity 400cst.
ARWRSWALIGRKQA-UHFFFAOYSA-N
methoxy-dimethyl-[methyl(diphenyl)silyl]oxysilane
121233058
1.07 g/mL
High temperature, Usable temp. range: 25 to 250°C (open system) and 25 to 315°C (closed system)
68083-14-7
100 g
MFCD00132673
14,8495
Miscible with aliphatic,aromatic hydrocarbons,alcohol and chlorinated hydrocarbons. Immiscible with water,methanol and ethylene glycol.
CO[Si](C)(C)O[Si](C)(C1=CC=CC=C1)C2=CC=CC=C2
302.52
1.505

Chemical Identifiers

68083-14-7
302.52
ARWRSWALIGRKQA-UHFFFAOYSA-N
121233058
CO[Si](C)(C)O[Si](C)(C1=CC=CC=C1)C2=CC=CC=C2
C16H22O2Si2
MFCD00132673
diphenyl-dimethylsiloxane copolymer, polydimethyl-diphenylsiloxane, viscosity 400cst.
methoxy-dimethyl-[methyl(diphenyl)silyl]oxysilane
Safety and Handling

Safety and Handling

RTECSNumber : VW3146875

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only