Binder™ Silicone-Free Door Gasket
Binder™ Silicone-Free Door Gasket for use with a range of Binder™ incubators, growth chambers, freezers, drying and heating chambers and climate chambers.
$886.52 - $1,240.73
Specifications
Material FKM Fluorocarbon
Type Door Gasket
View More Specs
Products 5
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity  
08111200 View Documents Binder™
80120500
VD 23, VDL 23 Each for $886.52
08111202 View Documents Binder™
80120502
VD 115, VDL 115 Each for $1,240.73
08111204 View Documents Binder™
80121317
VDL 53 Each for $1,062.63
08111203 View Documents Binder™
80121316
VDL 23 Each for $886.52
08111205 View Documents Binder™
80121318
VDL 115 Each for $1,240.73
Description
Description
  • Manufactured from FKM fluorocarbon
  • Temperature-resistant up to 200°C
Specifications
Specifications
FKM Fluorocarbon
Door Gasket