Alfa Aesar™ Silicon surface sensitizer, Gold chloride 0.5%
Manufacturer:  Alfa Aesar™ 44637MQ
 View more versions of this product
Catalog No. AA44637MQ
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Used for pretreatment of planar silicon devices when plating with Nickel plating solution products 44069 and 44070
Specifications
Specifications
Silicon surface sensitizer
1pt.
Gold chloride 0.5%
MFCD06412431
Documents