Please login to your online account to display your discounted pricing

Silicon Standard For ICP, 1000μg/mL in H2O/0.24 F, Assurance Grade, SPEX CertiPrep™


ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Quantity 125mL
Packaging Triple-leached LDPE bottle

Manufacturer: spex certiprep llc  PLSI92Y

 View more versions of this product

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

 Disclaimers

Product intended for laboratory use only

Catalog No. PLSI9-2Y

  • / Each
 

Description & Specifications

Chemical Identifiers

Packaging Triple-leached LDPE bottle
Quantity 125mL
DOT Information Nonhazardous
Recommended Storage Ambient
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Solvent or Matrix H2O/0.4% F
Purity Grade Assurance Grade
Certificate of Analysis Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
Chemical Name or Material Silicon Standard
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.H2O/tr. HF    1000μg/mL    PLSI9-2Y, -2T, -2X    10,000    PLSI9-3Y, -3XH2O    1000μg/mL    PLSI9A-2X    10,000    PLSI9A-3X
CAS Min % 0.1
CAS 16919-19-0
CAS Max % 0.1
CAS Min % 99.9
CAS 7732-18-5
CAS Max % 99.9

Specifications

Chemical Name or Material Silicon Standard
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Si1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Accessories

Ambient