Please login to your online account to display your discounted pricing

Silicon Standard, 1000mg/L in H2O/0.24 F (Assurance Grade), SPEX CertiPrep™


Quantity 30mL
Packaging Triple-leached LDPE bottle

Manufacturer: spex certiprep llc  SPXPLSI92M

 View more versions of this product

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Catalog No. PLSI92M

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 30mL
Purity Grade Assurance Grade
CAS 16919-19-0
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.H2O/tr. HF    1000μg/mL    PLSI9-2Y, -2T, -2X    10,000    PLSI9-
CAS Min % 0.1
CAS Max % 0.1
CAS 7732-18-5
CAS Min % 99.9
CAS Max % 99.9
Chemical Name or Material Silicon Standard
Health Hazard 1 Causes skin irritation, serious eye irritation. May cause respiratory irritation
DOT Information Nonhazardous
Packaging Triple-leached LDPE bottle
Recommended Storage Ambient
Solvent or Matrix H2O
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.

Specifications

Chemical Name or Material Silicon Standard
CAS 16919-19-0
CAS Min % 0.1
CAS Max % 0.1
CAS 7732-18-5
CAS Min % 99.9
CAS Max % 99.9
Purity Grade Assurance Grade

AnalyteConcentration
Si1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

GHS/OSHA Complaint

Accessories

Causes skin irritation, serious eye irritation. May cause respiratory irritation

Ambient