Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.999% (metals basis)

$589.54 - $589.54

Chemical Identifiers

Chemical Name or Material Silicon sputtering target
CAS 7440-21-3
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00085311
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40893KS

  SDS
alfa aesar™
40893KS
1 ea. Each for $589.54
Description & Specifications

Specifications

Quantity 1 ea.
Chemical Name or Material Silicon sputtering target
Odor Odorless
TSCA TSCA
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Assay 99.999%
Assay Percent Notes (metals basis)
MDL Number MFCD00085311
CAS 7440-21-3