Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.999% (metals basis)

$485.18 - $485.18

Chemical Identifiers

Chemical Name or Material Silicon sputtering target
CAS 7440-21-3
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00085311
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40891KS

  SDS
alfa aesar™
40891KS
1 ea. Each for $485.18
Description & Specifications

Specifications

Quantity 1 ea.
Chemical Name or Material Silicon sputtering target
CAS 7440-21-3
Name Note 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
MDL Number MFCD00085311
Odor Odorless
TSCA TSCA
Assay Percent Notes (metals basis)
Assay 99.999%