Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon sputtering target, 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick, 99.999% (metals basis)

Magnetron sputtering source

$293.82 - $293.82

Chemical Identifiers

Chemical Name or Material Silicon sputtering target
CAS 7440-21-3
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
MDL Number MFCD00085311
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40890KS

  SDS
alfa aesar™
40890KS
1 ea. Each for $293.82
Description & Specifications

Specifications

Quantity 1 ea.
Chemical Name or Material Silicon sputtering target
Odor Odorless
TSCA TSCA
Name Note 50.8mm (2.0 in.) dia. x 3.18mm (0.125 in.) thick
Assay 99.999%
Assay Percent Notes (metals basis)
MDL Number MFCD00085311
CAS 7440-21-3