Please login to your online account to display your discounted pricing

Silicon, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical


For atomic absorption. (1mL = 1mg Si) Solute: Sodium Silicate Solvent: Distilled Water

Quantity 500mL
Packaging Poly Bottle

Manufacturer: fisher scientific company  SS465500

 View more versions of this product

Catalog No. SS465-500

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Quantity 500mL
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
Color Colorless
pH 11.8
Chemical Name or Material Silicon, Reference Standard Solution
Concentration 990 to 1010ppm
ChemAlert Storage Symbol White
CAS 6834-92-0
Name Note 1000ppm ±1%
MDL Number MFCD00085311
CAS 7732-18-5
Packaging Poly Bottle

Specifications

Chemical Name or Material Silicon, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 6834-92-0
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00085311
Accessories
 

SDS & Safety Documents