Please login to your online account to display your discounted pricing

Silicon, Reference Standard Solution (1000ppm ±1%/Certified), Fisher Chemical

For atomic absorption. (1mL = 1mg Si) Solute: Sodium Silicate Solvent: Distilled Water

$84.76 - $115.53

Chemical Identifiers

Chemical Name or Material Silicon, Reference Standard Solution
Name Note 1000ppm ±1%
CAS 6834-92-0
CAS 7732-18-5
Purity Grade Certified Grade
MDL Number MFCD00085311
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SS465-100

  SDS
fisher scientific company
FLSS465100
Poly Bottle 100mL Each for $84.76
Case of 6 Each for $508.56

SS465-500

  SDS
fisher scientific company
SS465500
Poly Bottle 500mL Each for $115.53
Case of 6 Each for $693.18
Description & Specifications

Specifications

Purity Grade Certified Grade
Quantity 100mL
DOT Information DOT Class 8, : Corrosive
Physical Form Liquid
pH 11.8
Color Colorless
MDL Number MFCD00085311
CAS 7732-18-5
Concentration 990 to 1010ppm
Chemical Name or Material Silicon, Reference Standard Solution
CAS 6834-92-0
Packaging Poly Bottle
ChemAlert Storage Symbol White
Name Note 1000ppm ±1%
Safety and Handling