missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicon Powder, crystalline, APS≤50nm, 98%, , Alfa Aesar™

Form: S.A. 70 to 100m2/g

Manufacturer:  Thermo Fisher Scientific 4438409

 View more versions of this product

Catalog No. AA4438409



Specifications

Specifications

Silicon
98%
MFCD00085311
ccris 6599
[Si]
28.085
CHEBI:27573
2.33g/cm3
Air Sensitive
7440-21-3
Si
Powder
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
silicon
5461123
Crystalline Powder
Odorless
10g

Chemical Identifiers

7440-21-3
28.085
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
5461123
silicon
Si
MFCD00085311
ccris 6599
CHEBI:27573
[Si]
Safety and Handling

Safety and Handling

Take precautionary measures against static discharges.

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-130-8

RTECSNumber : VW0400000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Argon

Documents