missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon powder, crystalline, APS≈100nm, 99%, Plasma Synthesized

Manufacturer:  Alfa Aesar™ 4630804

 View more versions of this product

Catalog No. AA4630804



Specifications

Specifications

Silicon powder
Plasma Synthesized
APS ≈100nm
99%
MFCD00085311
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
silicon
5461123
Crystalline
2g
7440-21-3
Odorless
Si
UN1346
[Si]
28.085
CHEBI:27573

Chemical Identifiers

7440-21-3
28.085
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
CHEBI:27573
[Si]
Si
MFCD00085311
5461123
silicon
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P370+P378c
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: SILICON POWDER, AMORPHOUS

RTECSNumber : VW0400000

TSCA : TSCA

SDS Documents