missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon powder, crystalline, APS≈100nm, 99%, Plasma Synthesized

Manufacturer:  Alfa Aesar™ 4630809

 View more versions of this product

Catalog No. AA4630809



Specifications

Specifications

Silicon powder
Plasma Synthesized
APS ≈100nm
Si
UN1346
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
silicon
5461123
Crystalline
10g
7440-21-3
Odorless
MFCD00085311
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
[Si]
28.085
CHEBI:27573
99%

Chemical Identifiers

7440-21-3
28.085
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
5461123
silicon
Si
MFCD00085311
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831
CHEBI:27573
[Si]
Safety and Handling

Safety and Handling

GHS H Statement
H228
Flammable solid.

GHS P Statement
P210-P280-P240-P241-P370+P378c
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: SILICON POWDER, AMORPHOUS

RTECSNumber : VW0400000

TSCA : TSCA

SDS Documents