missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Silicon powder, crystalline, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01268109
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Silicon | |
Crystalline Powder | |
99.5% (Metals basis) | |
MFCD00085311 | |
XUIMIQQOPSSXEZ-UHFFFAOYSA-N | |
silicon | |
5461123 | |
Odorless |
7440-21-3 | |
10 g | |
Si | |
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 | |
[Si] | |
28.09 | |
CHEBI:27573 | |
Powder |
Chemical Identifiers
7440-21-3 | |
28.09 | |
XUIMIQQOPSSXEZ-UHFFFAOYSA-N | |
5461123 | |
silicon |
Si | |
MFCD00085311 | |
metal, silicone, defoamer s-10, flots 100sco, powder, dust, unii-z4152n8iui, polyeristalline powder, ccris 6599, ccris 6831 | |
CHEBI:27573 | |
[Si] |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
DOTInformation : DOT Class: 4.1, Packaging Group: III
EINECSNumber : 231-130-8
RTECSNumber : VW0400000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only