Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon powder, crystalline, -140 mesh, 98% (metals basis)

For plasma spraying

Chemical Identifiers

Chemical Name or Material Silicon powder
CAS 7440-21-3
MDL Number MFCD00085311
UN Number UN1346
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3968818

 
alfa aesar™
3968818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3968830

 
alfa aesar™
3968830
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Quantity 50g
Color Dark gray
Chemical Name or Material Silicon powder
Assay 98%
Assay Percent Notes (metals basis)
MDL Number MFCD00085311
CAS 7440-21-3
UN Number UN1346
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H228
Flammable solid.
Health Hazard 3 GHS P Statement
P210-P280-P240-P241-P370+P378c
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: SILICON POWDER, AMORPHOUS
Mesh Size −140 Mesh
Odor Odorless
TSCA TSCA
RTECS Number VW0400000
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P241-P370+P378c
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
Use explosion-proof electrical/ventilating/lighting/ equipment.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.

GHS H Statement
H228
Flammable solid.