Please login to your online account to display your discounted pricing

Silicon powder, crystalline, -100 mesh, 99.9% (metals basis), Alfa Aesar

$111.19 - $111.19

Chemical Identifiers

Chemical Name or Material Silicon
CAS 7440-21-3
Physical Form Crystalline Powder
Assay Percent Range 99.9% (Metals basis)
Molecular Formula Si
Formula Weight 28.0855
MDL Number MFCD00085311
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA0031122

  SDS
alfa aesar
0031122
100g Each for $111.19

AA0031136

  SDS
alfa aesar
0031136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 28.0855
Physical Form Crystalline Powder
Quantity 100g
DOT Information DOT Class: 4.1, Packaging Group: III
Density 2.33g/cm3
MDL Number MFCD00085311
CAS 7440-21-3
Chemical Name or Material Silicon
EINECS Number 231-130-8
Health Hazard 1 H228-H303: Flammable solid. May be harmful if swallowed.
Assay Percent Range 99.9% (Metals basis)
Appearance Powder
Odor Odorless
Molecular Formula Si
Safety and Handling

H228-H303: Flammable solid. May be harmful if swallowed.