missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicon powder, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4418530

 View more versions of this product

Catalog No. AA4418530



Specifications

Specifications

Silicon
7440-21-3
Si
Powder
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
silicon
5461123
Powder
250g
For analysis APS
99.9% (Metals basis)
MFCD00085311
ccris 6599
[Si]
28.085
CHEBI:27573
Odorless

Chemical Identifiers

7440-21-3
28.085
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
5461123
silicon
Si
MFCD00085311
ccris 6599
CHEBI:27573
[Si]
Safety and Handling

Safety and Handling

P210-P240-P241-P280-P370+P378q

H228

DOTInformation : DOT Class: 4.1, Packaging Group: III

EINECSNumber : 231-130-8

RTECSNumber : VW0400000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents