Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon, plasma standard solution, Specpure™, Si 10,000μg/mL

Chemical Identifiers

Chemical Name or Material Silicon plasma standard solution
Name Note Si 10000μg/mL
Purity Grade Specpure™
MDL Number MFCD00085311
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA38723AD

  SDS
alfa aesar™
38723AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA38723AE

  SDS
alfa aesar™
38723AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA38723AP

  SDS
alfa aesar™
38723AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Silicon plasma standard solution
Name Note Si 10000μg/mL
TSCA TSCA
Purity Grade Specpure™
Physical Form Liquid
MDL Number MFCD00085311
Quantity 50mL