Please login to your online account to display your discounted pricing

Silicon plasma standard solution, Alfa Aesar™, Specpure™

Chemical Identifiers

Chemical Name or Material Silicon
Name Note plasma standard solution
Physical Form Liquid
Molecular Formula Si in 5% HNO3/tr· HF
MDL Number MFCD00085311
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA13814AD

 
alfa aesar
13814AD
50mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA13814AP

 
alfa aesar
13814AP
500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 50mL
DOT Information DOT Class: 8, Packaging Group: III
Chemical Name or Material Silicon
Appearance Liquid
Name Note plasma standard solution
Physical Form Liquid
Molecular Formula Si in 5% HNO3/tr· HF
MDL Number MFCD00085311
Concentration or Composition (by Analyte or Components) Nitric acid: 5%; Hydrogen fluoride: 0.1%; Silicon: 0.1%; Water: 94.8%
Health Hazard 1 H314-H303-H313-H333: Causes severe skin burns and eye damage. May be harmful if swallowed. May be harmful in contact with skin. May be harmful if inhaled.
Vapor Pressure 23 hPa (17mm Hg) at 20°C
Safety and Handling

H314-H303-H313-H333: Causes severe skin burns and eye damage. May be harmful if swallowed. May be harmful in contact with skin. May be harmful if inhaled.