Please login to your online account to display your discounted pricing

Silicon oxide, Catalyst Support, High Surface Area, Alfa Aesar™

Form: S.A. 160m2/g

$38.86 - $384.23

Chemical Identifiers

Chemical Name or Material Silicon oxide
Name Note High Surface Area
MDL Number MFCD00011232
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4474122

  SDS
alfa aesar
4474122
100g Each for $38.86

AA4474136

  SDS
alfa aesar
4474136
500g Each for $125.07

AA44741A3

  SDS
alfa aesar
44741A3
2kg Each for $384.23
Description & Specifications

Specifications

Quantity 100g
Color White
Chemical Name or Material Silicon oxide
Odor Odorless
Name Note High Surface Area
Appearance White pellets
MDL Number MFCD00011232