missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicon Oxide, Catalyst Support, Alfa Aesar™

Form: S.A. 250m2/g

Manufacturer:  Alfa Aesar 4474036

 View more versions of this product

Catalog No. AA4474036



Specifications

Specifications

Silicon oxide
O2Si
White pellets
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
Odorless
White
7631-86-9
MFCD00011232
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
O=[Si]=O
60.083
CHEBI:30563
3.2mm (0.125 in.)
500g

Chemical Identifiers

7631-86-9
60.083
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
dioxosilane
O2Si
MFCD00011232
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
CHEBI:30563
O=[Si]=O
SDS Documents