Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon, Oil based standard solution, Specpure™, Si 1000μg/g

Chemical Identifiers

Chemical Name or Material Silicon Oil based standard solution
Name Note Si 1000μg/g
Purity Grade Specpure™
MDL Number MFCD00085311
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1420918

  SDS
alfa aesar™
1420918
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Purity Grade Specpure™
MDL Number MFCD00085311
Quantity 50g
Chemical Name or Material Silicon Oil based standard solution
Name Note Si 1000μg/g