missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon lump, 10cm (3.9 in.) & down, 98.4% (metals basis)

Chemical Identifiers

CAS 7440-21-3
Molecular Formula Si
Molecular Weight (g/mol) 28.085
MDL Number MFCD00085311
InChI Key XUIMIQQOPSSXEZ-UHFFFAOYSA-N
Synonym ccris 6599, ccris 6831, defoamer s-10, dust, flots 100sco, metal, polyeristalline powder, powder, silicone, unii-z4152n8iui
PubChem CID 5461123
ChEBI CHEBI:27573
IUPAC Name silicon
SMILES [Si]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA10335A1
View Documents
Alfa Aesar™
10335A1
1 kg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA10335A7
View Documents
Alfa Aesar™
10335A7
5 kg N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

7440-21-3
28.085
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
5461123
silicon
Si
MFCD00085311
ccris 6599, ccris 6831, defoamer s-10, dust, flots 100sco, metal, polyeristalline powder, powder, silicone, unii-z4152n8iui
CHEBI:27573
[Si]

Specifications

1 kg
Si
ccris 6599, ccris 6831, defoamer s-10, dust, flots 100sco, metal, polyeristalline powder, powder, silicone, unii-z4152n8iui
[Si]
Odorless
5461123
Lumps
98.4%
(metals basis)
7440-21-3
MFCD00085311
XUIMIQQOPSSXEZ-UHFFFAOYSA-N
silicon
28.085
CHEBI:27573
10cm (3.9 in.) & down
Silicon lump
Safety and Handling

Safety and Handling

EINECSNumber : 231-130-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS