Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon(IV) oxide sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.995% (metals basis)

Chemical Identifiers

Chemical Name or Material Silicon(IV) oxide sputtering target
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Formula Weight 60.09
MDL Number MFCD00011232
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41099KS

  SDS
alfa aesar™
41099KS
1 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 60.09
Chemical Name or Material Silicon(IV) oxide sputtering target
TSCA TSCA
Name Note 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick
Assay 99.995%
Assay Percent Notes (metals basis)
MDL Number MFCD00011232
Quantity 1 each