Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon(IV) oxide, powder, 0.5 micron, 99.9%


Quantity 100g

Manufacturer: alfa aesar™  L1698522

 View more versions of this product

Catalog No. AAL1698522

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 60.09
Chemical Name or Material Silicon(IV) oxide
Health Hazard 3 GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 99.9%
CAS 7631-86-9
Physical Form Powder
Molecular Formula SiO2
Boiling Point 2230°C
MDL Number MFCD00011232
Health Hazard 2 GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.
Quantity 100g
Health Hazard 1 Warning
Melting Point 1710°C
Name Note 0.5 micron
TSCA TSCA
Merck Index 14,8493
Odor Odorless

Specifications

Chemical Name or Material Silicon(IV) oxide
CAS 7631-86-9
Name Note 0.5 micron
Molecular Formula SiO2
Formula Weight 60.09
MDL Number MFCD00011232
Merck Index 14,8493

Applications
Silicon(IV) oxide is used in the manufacture of glass for windows, drinking glasses, beverage bottles and many other materials. Optical fibers for telecommunication are made from silica. It is also used as a raw material for many ceramics such as earthenware, stoneware and porcelain. In addition, silica is a common additive in the production of foods and pharmaceutical products.

Solubility
Insoluble in water.

Notes
It is a very stable compound of silicon.

Accessories

Warning

GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.

SDS & Safety Documents