missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon(IV) oxide, amorphous fumed, S.A. 350-420m2/g

Manufacturer:  Alfa Aesar™ 89376A1

 View more versions of this product

Catalog No. AA89376A1



Specifications

Specifications

Silicon (IV) oxide
7631-86-9
MFCD00011232
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
O=[Si]=O
60.083
CHEBI:30563
Powder
−325 Mesh
1.46
2230°C
1710°C
Amorphous fumed
O2Si
14,8493
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
60.09
2.2g/mL
Odorless
350 to 420m2/g
White
1kg

Chemical Identifiers

7631-86-9
60.083
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
dioxosilane
O2Si
MFCD00011232
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
CHEBI:30563
O=[Si]=O
Safety and Handling

Safety and Handling

GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents