Please login to your online account to display your discounted pricing

Silicon(IV) oxide, Amorphous fumed, S.A. 300 to 350m2/g, Alfa Aesar™

For Adhesives, Coatings, Cosmetics

$29.24 - $146.69

Chemical Identifiers

Chemical Name or Material Silicon(IV) oxide
CAS 7631-86-9
Physical Form Powder
Molecular Formula O2Si
Formula Weight 60.09
MDL Number MFCD00011232
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4274118

  SDS
alfa aesar
4274118
50g Each for $29.24

AA4274130

  SDS
alfa aesar
4274130
250g Each for $93.43

AA42741A1

  SDS
alfa aesar
42741A1
1kg Each for $146.69
Description & Specifications

Specifications

Formula Weight 60.09
Quantity 50g
Color White
Density 2.2g/cm3 at 20°C
Chemical Name or Material Silicon(IV) oxide
EINECS Number 231-545-4
Odor Odorless
Physical Form Powder
Molecular Formula O2Si
MDL Number MFCD00011232
CAS 7631-86-9
Surface Area or Specific Surface S.A.:300 to 350m2/g
Health Hazard 1 H373: May cause damage to organs through prolonged or repeated exposure.
Health Hazard 2 Wear suitable protective clothing.

S.A. 300 to 350m2/g

Safety and Handling

H373: May cause damage to organs through prolonged or repeated exposure.

Wear suitable protective clothing.