missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicon(IV) oxide, Amorphous, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4478114

 View more versions of this product

Catalog No. AA4478114



Specifications

Specifications

Silicon(IV) oxide
99.9% (metals basis)
MFCD00011232
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
60.09
Odorless
25g
7631-86-9
O2Si
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
O=[Si]=O
60.083
CHEBI:30563
Powder
White

Chemical Identifiers

7631-86-9
60.083
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
dioxosilane
O2Si
MFCD00011232
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
CHEBI:30563
O=[Si]=O
Safety and Handling

Safety and Handling

Wear suitable protective clothing.

H373: May cause damage to organs through prolonged or repeated exposure.

EINECSNumber : 231-545-4

SDS Documents