Please login to your online account to display your discounted pricing

Silicon(IV) oxide, 99.99% (metals basis), 1 to 3mm fused lump, Alfa Aesar™

$220.12 - $220.12

Chemical Identifiers

Chemical Name or Material Silicon(IV) oxide
CAS 7631-86-9
Physical Form 1 To 3mm Fused Lump
Molecular Formula SiO2
Formula Weight 60.09
MDL Number MFCD00011232
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4373614

 
alfa aesar
4373614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4373622

 
alfa aesar
4373622
100g Each for $220.12
Description & Specifications

Specifications

Physical Form 1 To 3mm Fused Lump
Formula Weight 60.09
Quantity 25g
Color White
Chemical Name or Material Silicon(IV) oxide
Odor Odorless
Molecular Formula SiO2
MDL Number MFCD00011232
CAS 7631-86-9