missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Silicon(IV) oxide, 99.95% (metals basis)

Manufacturer:  Alfa Aesar™ 4709822

 View more versions of this product

Catalog No. AA4709822



Specifications

Specifications

Silicon (IV) oxide
(metals basis)
MFCD00011232
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
O=[Si]=O
60.083
CHEBI:30563
Powder
2.2g/mL
Odorless
White
100g
7631-86-9
O2Si
14,8493
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
60.09
99.95%
−40 Mesh
2230°C
1710°C

Chemical Identifiers

7631-86-9
60.083
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
dioxosilane
O2Si
MFCD00011232
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
CHEBI:30563
O=[Si]=O
Safety and Handling

Safety and Handling

GHS H Statement
H373
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P314-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical advice/attention if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

SDS Documents