missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicon(IV) oxide, 99.8% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 8970930

 View more versions of this product

Catalog No. AA8970930



Specifications

Specifications

Silicon(IV) oxide
99.8% (metals basis)
MFCD00011232
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
60.09
2.2 to 2.6g/mL
7631-86-9
O2Si
silicates crystalline silica :graphite natural, silica flour powdered crystalline silica, silica amorphous fumed cryst.-free, silicon dioxide chemically prepared, amorphous silica: pyrogenic fumed, cristobalite silica crystalline, epa pesticide chemical code 072605, silica crystalline-cristobalite, silica crystalline: cristobalite, synthetic amorphous silica fumed
O=[Si]=O
60.083
CHEBI:30563
-325 Mesh Fused Amorphous Powder, S.A. 5m2/g
250g

Chemical Identifiers

7631-86-9
60.083
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
dioxosilane
O2Si
MFCD00011232
silicates crystalline silica :graphite natural, silica flour powdered crystalline silica, silica amorphous fumed cryst.-free, silicon dioxide chemically prepared, amorphous silica: pyrogenic fumed, cristobalite silica crystalline, epa pesticide chemical code 072605, silica crystalline-cristobalite, silica crystalline: cristobalite, synthetic amorphous silica fumed
CHEBI:30563
O=[Si]=O
Safety and Handling

Safety and Handling

EINECSNumber : 231-545-4

SDS Documents