missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Silicon(IV) oxide, 99.5% (metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 88316A1

 View more versions of this product

Catalog No. AA88316A1



Specifications

Specifications

Silicon(IV) oxide
99.5% (metals basis)
MFCD00011232
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
dioxosilane
24261
60.09
Odorless
1kg
14808-60-7
O2Si
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
O=[Si]=O
60.083
CHEBI:30563
Powder
Brown to White

Chemical Identifiers

14808-60-7
60.083
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
dioxosilane
O2Si
MFCD00011232
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
CHEBI:30563
O=[Si]=O
Safety and Handling

Safety and Handling

H350-H373: May cause cancer. May cause damage to organs through prolonged or repeated exposure.

EINECSNumber : 238-878-4

SDS Documents